QuantiGene™ Singleplex Assay Kit
Invitrogen™

QuantiGene™ Singleplex Assay Kit

QuantiGene Singleplex assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates, tissue homogenates or purified RNA.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
QS00122 Plates
QS001310 Plates
QS001650 Plates
카탈로그 번호 QS0012
제품 가격(KRW)
1,720,000
온라인 행사
Ends: 30-Sep-2026
1,911,000
할인액 191,000 (10%)
Each
수량:
2 Plates
제품 가격(KRW)
1,720,000
온라인 행사
Ends: 30-Sep-2026
1,911,000
할인액 191,000 (10%)
Each
QuantiGene Singleplex assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates, tissue homogenates or purified RNA. They utilize branched DNA technology for signal amplification, and signal is detected using a standard luminometer. The QuantiGene Singleplex Assay Kit contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates. Sample processing kits and target-specific probe sets are sold separately.

The QuantiGene Singleplex Assay is comprised of three modules:

  • QuantiGene Singleplex Assay Kit
  • QuantiGene Singleplex target-specific probe sets (sold separately)
  • QuantiGene Sample Processing Kit (sold separately) (not required for purified RNA samples)

Features of the QuantiGene Singleplex Assay include:

  • Direct from lysate quantitation using signal amplification—no RNA or DNA purification, reverse transcription, or PCR amplification needed
  • Precisely quantify subtle differences in gene expression as small as 10%
  • Simple workflow features lyse-and-go sample preparation, easily automatable ELISA-like protocol, and readout using any standard luminometer
  • Analyze any sample type: cultured cells; whole blood or dried blood spots; fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues; plants; bacteria

QuantiGene Singleplex assays bring the power of branched DNA (bDNA) technology to basic and pre-clinical research. The bDNA assay is a sandwich nucleic acid hybridization method that uses bDNA molecules to amplify signal from captured target RNA. bDNA technology forms the basis of the FDA-approved clinical diagnostic VERSANT 3.0 assays for HIV, HCV, and HBV viral load that are offered commercially by Siemens and have been in use for over a decade. Another advantage of bDNA assays is that RNA is measured directly from the sample source, without RNA purification or enzymatic manipulation, thereby avoiding inefficiencies and variability introduced by or errors inherent to these processes.

Each capture plate in this assay kit is packaged individually in a sealed foil pouch.

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(애플리케이션)QuantiGene Singleplex Probe Set
용도(장비)Luminometer
제품라인QuantiGene Singleplex
검출 방법Chemiluminescence
형식96-well
제품 유형Singleplex Assay Kit
수량2 Plates
Unit SizeEach
구성 및 보관

• Blocking Reagent (400 μL, store at 2°C to 8°C)
• Pre-Amplifier Solution (23 mL, store at 2°C to 8°C)
• Amplifier Solution (23 mL, store at 2°C to 8°C)
• Capture Plate (2 ea, packaged individually, store at 2°C to 8°C)
• Label Probe Solution (23 mL, store at 2°C to 8°C)
• Substrate (24 mL, store at 2°C to 8°C)
• Lysis Mixture (18 mL, store at 15–30°C)
• Wash Buffer Component 1 (3 mL, store at 15–30°C)
• Wash Buffer Component 2 (5 mL, store at 15–30°C)
• Plate Seals Day 1 (2 ea, store at 15–30°C)
• Plate Seals Day 2 (10 ea, store at 15–30°C)

자주 묻는 질문(FAQ)

How does the QuantiGene Singleplex Assay Kit (Cat. No. QS0011) differ from the new kit (Cat. Nos. QS0012, QS0013, QS0014, QS0016)?

The QuantiGene Singleplex Assay Kit has been reformulated such that the Pre-Amplifier Solution and Amplifier Solution now come pre-diluted, which streamlines the workflow, and also these solutions can be shipped and stored at 2-8 degrees C now instead of -20 degrees C. The Blocking Reagent is also now shipped and stored at 2-8 degrees C. Hence, the QuantiGene Singleplex Assay Kit (Cat. No. QS0011) has been discontinued and replaced with the new kit (Cat. Nos. QS0012, QS0013, QS0014, QS0016).