QuantiGene™ Singleplex Assay Kit
Invitrogen™

QuantiGene™ Singleplex Assay Kit

QuantiGene Singleplex assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates, tissue homogenates or purified RNA.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
QS001310 Platten
QS00122 Platten
QS001650 Platten
Katalognummer QS0013
Preis (EUR)
6.100,00
Each
Menge:
10 Platten
Preis (EUR)
6.100,00
Each
QuantiGene Singleplex assays are hybridization-based assays for the direct quantitation of RNA, DNA, or microRNA transcripts directly from cell lysates, tissue homogenates or purified RNA. They utilize branched DNA technology for signal amplification, and signal is detected using a standard luminometer. The QuantiGene Singleplex Assay Kit contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates. Sample processing kits and target-specific probe sets are sold separately.

The QuantiGene Singleplex Assay is comprised of three modules:

  • QuantiGene Singleplex Assay Kit
  • QuantiGene Singleplex target-specific probe sets (sold separately)
  • QuantiGene Sample Processing Kit (sold separately) (not required for purified RNA samples)

Features of the QuantiGene Singleplex Assay include:

  • Direct from lysate quantitation using signal amplification—no RNA or DNA purification, reverse transcription, or PCR amplification needed
  • Precisely quantify subtle differences in gene expression as small as 10%
  • Simple workflow features lyse-and-go sample preparation, easily automatable ELISA-like protocol, and readout using any standard luminometer
  • Analyze any sample type: cultured cells; whole blood or dried blood spots; fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues; plants; bacteria

QuantiGene Singleplex assays bring the power of branched DNA (bDNA) technology to basic and pre-clinical research. The bDNA assay is a sandwich nucleic acid hybridization method that uses bDNA molecules to amplify signal from captured target RNA. bDNA technology forms the basis of the FDA-approved clinical diagnostic VERSANT 3.0 assays for HIV, HCV, and HBV viral load that are offered commercially by Siemens and have been in use for over a decade. Another advantage of bDNA assays is that RNA is measured directly from the sample source, without RNA purification or enzymatic manipulation, thereby avoiding inefficiencies and variability introduced by or errors inherent to these processes.

Each capture plate in this assay kit is packaged individually in a sealed foil pouch.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Zur Verwendung mit (Anwendung)QuantiGene Singleplex Probe Set
Zur Verwendung mit (Geräte)Luminometer
ProduktlinieQuantiGene Singleplex
NachweisverfahrenChemiluminescence
Format96-Well
ProdukttypSingleplex Assay Kit
Menge10 Platten
Unit SizeEach
Inhalt und Lagerung

• Blocking Reagent (1.8 mL, store at 2°C to 8°C)

• Pre-Amplifier Solution (120 mL, store at 2°C to 8°C)
• Amplifier Solution (120 mL, store at 2°C to 8°C)
• Capture Plate (10 ea, packaged individually, store at 2°C to 8°C)
• Label Probe Solution (120 mL, store at 2°C to 8°C)
• Substrate (120 mL, store at 2°C to 8°C)
• Lysis Mixture (93 mL, store at 15–30°C)
• Wash Buffer Component 1 (15 mL, store at 15–30°C)
• Wash Buffer Component 2 (25 mL, store at 15–30°C)
• Plate Seals Day 1 (10 ea, store at 15–30°C)
• Plate Seals Day 2 (50 ea, store at 15–30°C)

Häufig gestellte Fragen (FAQ)

How does the QuantiGene Singleplex Assay Kit (Cat. No. QS0011) differ from the new kit (Cat. Nos. QS0012, QS0013, QS0014, QS0016)?

The QuantiGene Singleplex Assay Kit has been reformulated such that the Pre-Amplifier Solution and Amplifier Solution now come pre-diluted, which streamlines the workflow, and also these solutions can be shipped and stored at 2-8 degrees C now instead of -20 degrees C. The Blocking Reagent is also now shipped and stored at 2-8 degrees C. Hence, the QuantiGene Singleplex Assay Kit (Cat. No. QS0011) has been discontinued and replaced with the new kit (Cat. Nos. QS0012, QS0013, QS0014, QS0016).