Search
Search

| Katalognummer | Menge |
|---|---|
| QS0016 | 50 Platten |
| QS0012 | 2 Platten |
| QS0013 | 10 Platten |
The QuantiGene Singleplex Assay is comprised of three modules:
Features of the QuantiGene Singleplex Assay include:
QuantiGene Singleplex assays bring the power of branched DNA (bDNA) technology to basic and pre-clinical research. The bDNA assay is a sandwich nucleic acid hybridization method that uses bDNA molecules to amplify signal from captured target RNA. bDNA technology forms the basis of the FDA-approved clinical diagnostic VERSANT 3.0 assays for HIV, HCV, and HBV viral load that are offered commercially by Siemens and have been in use for over a decade. Another advantage of bDNA assays is that RNA is measured directly from the sample source, without RNA purification or enzymatic manipulation, thereby avoiding inefficiencies and variability introduced by or errors inherent to these processes.
Each capture plate in this assay kit is packaged individually in a sealed foil pouch.
• Blocking Reagent (10 mL, store at 2°C to 8°C)
• Pre-Amplifier Solution (550 mL, store at 2°C to 8°C)
• Amplifier Solution(550 mL, store at 2°C to 8°C)
• Capture Plate (50 ea, packaged individually, store at 2°C to 8°C)
• Label Probe Solution (550 mL, store at 2°C to 8°C)
• Substrate (530 mL, store at 2°C to 8°C)
• Lysis Mixture (410 mL, store at 15–30°C)
• Wash Buffer Component 1 (75 mL, store at 15–30°C)
• Wash Buffer Component 2 (125 mL, store at 15–30°C)
• Plate Seals Day 1 (50 ea, store at 15–30°C)
• Plate Seals Day 2 (210 ea, store at 15–30°C)