Search
| Número de catálogo | Cantidad |
|---|---|
| QS0639 | 10 μg |
Universal Human Reference RNA is an RNA extract used as a positive control in QuantiGene assays. The Universal Human Reference RNA is composed of total RNA from 10 human cell lines:
Equal quantities of DNase-treated total RNA from each cell line were pooled to make the universal control.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.