Universal Mouse Reference RNA for QuantiGene Assays
Invitrogen™
Universal Mouse Reference RNA for QuantiGene Assays
Universal Mouse Reference RNA is an RNA extract used as a positive control in QuantiGene assays.
Have Questions?
Catalog NumberQuantity
QS064010 μg
Catalog number QS0640
Price (CLP)
-
Quantity:
10 μg
Request bulk or custom format
Universal Mouse Reference RNA is an RNA extract used as a positive control in QuantiGene assays. The Universal Mouse Reference RNA is composed of total RNA from 11 mouse cell lines:
  • Embryo
  • Embryo, fibroblast
  • Kidney
  • Liver, hepatocyte
  • Lung, alveolar macrophage
  • B-lymphocyte
  • T-lymphocyte (thymus)
  • Mammary gland
  • Muscle myoblast
  • Skin
  • Testis
Equal quantities of DNase-treated total RNA from each cell line were pooled to make the universal control.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Nucleic Acid Quantitation
Product TypeReference RNA
Quantity10 μg
Sample SourceCell Line
Shipping ConditionShipped at 2°C to 8°C
Sample TypeRNA
SpeciesMouse
Unit SizeEach