Horizontal Air Flow Ovens, 220V, 1.6 cubic feet - FAQs
Horizontal Air Flow Ovens, 220V, 1.6 cubic feet - FAQs
View additional product information for Horizontal Air Flow Ovens, 220V, 1.6 cubic feet - FAQs (QS0712)
1 product FAQs found
To perform ViewRNA assays, which incubator oven should I use, Cat. No. QS0704 or QS0712?
Cat. No. QS0704 is a 120 V unit, for use in US/Canada region (https://www.thermofisher.com/order/catalog/product/QS0704).
Cat. No. QS0712 is a 220 V unit, for use in European region (https://www.thermofisher.com/order/catalog/product/QS0712).