Product Image
Invitrogen™
ViewRNA™ Temperature Validation Kit
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy. This kit may be used forRead more
Have Questions?
Catalog NumberQuantity
QV05231 Kit
Catalog number QV0523
Price (USD)
483.65
Online exclusive
532.00
Save 48.35
Each
Quantity:
1 Kit
Price (USD)
483.65
Online exclusive
532.00
Save 48.35
Each
The ViewRNA Temperature Validation Kit uses a calibrated digital thermometer to assess temperature accuracy. This kit may be used for any ViewRNA Assay to evaluate the temperature of incubators or ThermoBrite systems.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Kit
TypeTemperature Validation Kit
Unit SizeEach
Frequently asked questions (FAQs)
How do I calibrate my incubator oven for ViewRNA assays?
We recommend using the ViewRNA Temperature Validation Kit (Cat. No. QV0523) for verification and calibration of the incubator oven to a stable temperature. It may also be used to evaluate the temperature of ThermoBrite systems. The ViewRNA Temperature Validation Kit comes with a slide connected to a thermal probe attached to a calibrated digital thermometer that assesses the temperature inside the oven in a continuous manner. For ViewRNA assays, the incubator oven should be set at 40 ± 1 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.