Aminoallyl-dUTP Solution (10 mM)
Thermo Scientific™

Aminoallyl-dUTP Solution (10 mM)

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.Aminoallyl-dUTPRead more
Have Questions?
Catalog NumberQuantity
R0091100 μL
Catalog number R0091
Price (USD)
126.65
Online exclusive
139.00
Save 12.35
Each
Quantity:
100 μL
Request bulk or custom format
Price (USD)
126.65
Online exclusive
139.00
Save 12.35
Each

Thermo Scientific Aminoallyl-dUTP (5-[3-aminoallyl]-2'-deoxyuridine-5'-triphosphate) is supplied as 10 mM and 50 mM aqueous solutions titrated to pH 7.0 with NaOH.

Aminoallyl-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
• Taq DNA polymerase
• phi29 DNA Polymerase
• Klenow Fragment, Klenow Fragment, exo-
• DNA Polymerase I

The resulting amine-containing DNA can be subsequently labeled with any amine-reactive fluorescent dye, biotin, or hapten.

Applications
• Enzymatic indirect non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random-primed labeling
• Primer extension

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling MethodDirect Labeling
PolymeraseDNA Polymerase
Quantity100 μL
Shipping ConditionDry Ice
Concentration10 mM
For Use With (Application)cDNA Synthesis, Non-Radioactive Labeling
FormLiquid
Label or DyeAminoallyl
Unit SizeEach
Contents & Storage
Aminoallyl-dUTP Solution, 10 mM (100 μL)

Store at –20°C.