Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Inhibit gram-positive microorganisms and facultative anaerobic bacteria and select for gram-negative bacilli with Thermo Scientific™ Anaerobic KV Blood Agar [The Centers for Disease Control and Prevention (CDC) formulation] with kanamycin and vancomycin.
Formulated by V.R. Dowell and T.M Hawkins at the CDC in Atlanta Georgia1, Anaerobic Blood Agar (CDC) supports the growth and typical pigmentation of fastidious, slow-growing and other clinically important anaerobes.
Description | Anaerobic KV Blood Agar |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | Anaerobic KV Blood Agar |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 100/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01042 | Each | 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01043 | Each | 100/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Isolate and cultivate anaerobic organisms, including fastidious strains using the solid medium Remel Anaerobic KV Blood Agar (CDC formulation) with kanamycin and vancomycin.
Anaerobic Blood Agar (CDC) base contains peptones that supply nitrogenous substances and amino acids necessary for the growth of anaerobic bacteria. Yeast extract provides B-complex vitamins and serves as a growth enhancer. Hemin, vitamin K and sheep blood stimulate the growth of anaerobes. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. Kanamycin and vancomycin are antibiotics which inhibit gram-positive organisms and facultative anaerobic bacteria and select for gram-negative bacilli.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.