Thermo Scientific™
Remel™ Anaerobic Neomycin Blood Agar
 Related applications:
Clinical Microbiology
Thermo Scientific™ Remel Anaerobic Neomycin Blood Agar (CDC formulation) is used for the isolation of anaerobic microorganisms such as Bacteroides and Clostridium spp.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Neomycin inhibits gram-negative Enterobacteriaceae
Documents
Manuals & protocols