Anaerobic Reducible Blood Agar with colistin and nalidixic acid (CNA)
Product Image
Thermo Scientific™
Anaerobic Reducible Blood Agar with colistin and nalidixic acid (CNA)
Cultivate and isolate fastidious anaerobic bacteria with this selective, ready-to-use solid medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R01063100/Pk.
R0106410/Pk.
Catalog number R01063
Price (USD)
-
Request A Quote
Quantity:
100/Pk.
Cultivate and isolate fastidious anaerobic bacteria with Thermo Scientific™ Remel™ Anaerobic Reducible Blood Agar.
The addition of cysteine, palladium chloride, and dithiothreitol makes the medium reducible. Reducible medium improves the recovery of anaerobes by lowering the oxidation – reduction potential of the medium. Kanamycin and vancomycin are added as selective agents for recovery of Gram-negative anaerobes, especially Bacteriodes spp
Anaerobic Reducible Blood Agar is a solid media recommended for use in qualitative procedures for the primary isolation and cultivation of fastidious anaerobic bacteria.
  • Selective: Kanamycin and vancomycin antibiotics inhibit gram-positive organisms and facultative anaerobic bacteria. Neomycin stimulates the growth of gram-negative anaerobes, especially Bacteriodes spp. Colistin and nalidixic inhibit the growth of most gram-negative bacilli.
  • Ready-to-use: Convenience of prepared media.
Anaerobic Reducible Blood Agar is a nutritious medium containing peptone, hemin, vitamin K and sheep blood, which supply anaerobic organisms with required growth factors. Colistin and nalidixic inhibit the growth of most gram-negative bacilli. Kanamycin and vancomycin inhibit gram-positive organisms and facultative anaerobic bacteria. Neomycin facilitates the isolation of anaerobic organisms, such as Bacteriodes and Clostridium spp. and inhibits enteric gram-negative bacilli. Laked blood is added to enhance pigment formation by anaerobic bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
DescriptionAnaerobic Reducible; CNA Agar Base
Format12mm x 85mm Monoplate
Quantity100/Pk.
Product TypeAgar
Unit SizeEach