Bacteroides Fragilis Isolation Agar
Thermo Scientific™
Bacteroides Fragilis Isolation Agar
Isolate and presumptively identify Bacteroides fragilis group from clinical specimens with this selective, ready-to-use medium.
Have Questions?
Catalog NumberdimFormat
R01104
Catalog number R01104
Price (MXN)
-
Isolate and presumptively identify Bacteroides fragilis group from clinical specimens with the selective, differential Thermo Scientific™ Remel™ Bacteroides Fragilis Isolation Agar. The selectivity of this medium is provided by the addition of gentamicin and bile. Differentiation is facilitated by the hydrolysis of esculin to esculitin by the Bacteroides fragilis group. Ferric ammonium citrate is added to the medium to react with esculetin and produce a brown-black complex surrounding the colony.
Bacteroides fragilis is an obligately anaerobic, Gram-negative, rod-shaped bacterium. It is part of the normal microbiota of the human colon and is generally commensal, but can cause infection if displaced into the bloodstream or surrounding tissue following surgery, disease, or trauma.
Bacteroides Fragilis Isolation Agar is a selective and differential medium for recovery and presumptive identification of Bacteroides fragilis group.
  • Easy-to-read: Bacteroides fragilis colonies are easy to read due to brown-black complex surrounding the colonies.
  • Selective: gentamicin and bile make the medium selective for Bacteroides fragilis. Gentamicin inhibits most facultative anaerobes and bile inhibits anaerobic gram-negative bacilli with the exception of Bacteroides fragilis group and other Bacteroids and Fusobacteria.
  • Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
General References:
  1. Food and Drug Administration. 1995. Bacteriological Analytical Manual. 8th ed. AOAC International. Gaithesburg, MD.
  2. Dowens, F.P. and K. Ito 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th e. APHA, Washington, D.c.
Specifications
DescriptionBacteroides Fragilis Isolation Agar
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach