Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious base medium Thermo Scientific™ Remel™ Blood Heart Infusion (BHI) Agar with Sheep Blood, Chloramphenicol and Cycloheximide1. Isolate fastidious fungi and grow Actinomyces2 using BHI Agar with 5-10% sheep blood. Pathogenic fungi and Nocardia asteroides can be isolated using optimal concentrations of chloramphenicol3. Cycloheximide inhibits certain Aspergillus spp. and zygomycetes.
Description | BHI Agar w/Sheep Blood, Chloramphenicol, Cycloheximide |
Format | Monoplate Deep Pour |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Format | 20mm x 113mm Tube |
Type | Agar |
Description | BHI Agar w/Sheep Blood, Chloramphenicol, Cycloheximide |
Product Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Format | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01142 | Each | Monoplate Deep Pour | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08073 | Each | 20mm x 113mm Tube | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Use BHI Agar with Sheep Blood, Chloramphenicol and Cycloheximide1 for the cultivation of a wide variety of microorganisms, including pathogenic fungi.
BHI Agar w/Sheep Blood contains brain heart infusion, peptone, and dextrose, which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.