Thermo Scientific™
Remel™ BHI Agar with Vancomycin
Catalog number: R01176
Thermo Scientific™
Remel™ BHI Agar with Vancomycin
Catalog number: R01176
Detect vancomycin-resistant enterococci with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar with Vancomycin. Brain Heart Infusion (BHI) Agar with Vancomycin is used for screening of enterococci resistant to vancomycin and is recommended by the Clinical and Laboratory Standards Institute (CLSI) as a reliable method for the detection of all phenotypes of vancomycin-resistant enterococci1,2.
Catalog Number
R01176
Unit Size
Each
dimFormat
Price (USD)
Full specifications
DescriptionBHI Agar w/Vancomycin 6mcg/mL; Monoplate
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizedimFormatPrice (USD)
R01176Full specifications
EachRequest A Quote
DescriptionBHI Agar w/Vancomycin 6mcg/mL; Monoplate
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Showing 1 of 1
Vancomycin-resistant enterococci were first recognized in Europe and the United States in the late 1980s. Since then they are now being recovered with increasing frequency3. Various phenotypes responsible for acquired resistance in enterococci have been described with high to low vancomycin resistance and cross-resistance to teicoplanin4. Detection has been difficult due to failure of commonly used susceptibility methods to identify those isolates with low-level resistance5.
An Agar screen test with BHI Agar supplemented with 6µg/ml of vancomycin has been recommended by the Clinical and Laboratory Standards Institute (CLSI) as a reliable method for the detection of all phenotypes of vancomycin-resistant enterococci1,2.
  • Recommended by the CLSI as a reliable method for the detection of all phenotypes of vancomycin-resistant enterococci1,2
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
1. Clinical and Laboratory Standards Institute (CLSI). 2009. Methods for Dilution Antimicrobial Susceptibility Tests for Bacteria that Grow Aerobically; Approved Standards, 8th Edition, m74-A8, CLSI. Wayne, PA.
2. Clinical and Laboratory Standards Institute (CLSI). 2010. Performance Standards for Antimicrobial Susceptibility Testing; 20th Informational Supplement. Mq00-S20, CLSI, Wayne, PA.
3. Leclercq, R., E. Derlot, J. Duval, and P. Courvalin. 1988. N.Eng.J.Med. 319:157 -161.
4. Clark, N.C., R.C. Cooksey, B.C. Hill, J.M. Swenson, and F.C. Tenover. 1993. Antimicrobial Agents and Chermother. 37:2311-2317.
5. Wiley, B.M., B.N. Kreiswirth, A.E. Simor, G. Williams, S.R. Scriever, A. Phillips, and D.E. Low 1992 .J.Clin.Micribiol. 30:1621 -1624.