Product Image
Thermo Scientific™

Blood Agar with 5% Rabbit Blood

Cultivate Haemophilus species with this ready prepared medium.

Have Questions?
Catalog NumberFormat
R0121012mm x 85mm Monoplate
Catalog number R01210
Price (USD)
-
Format:
12mm x 85mm Monoplate

Cultivate Haemophilus species with Thermo Scientific™ Remel™ Blood Agar with 5% Rabbit Blood. Haemophilus spp. requires hemin (X factor) and NAD (V factor) for growth, both of which are supplied by Rabbit blood. Horse blood does not contain V factor -inactivating enzymes and supplies X and V factors, one or both of which are required for the growth of Haemophilus spp. Rabbit blood also allows for differentiation of hemolytic and nonhemolytic species of Haemophilus.

Members of  the genus Haemophilus are fastidious Gram-negative bacilli that commonly cause infections of upper respiratory tract. Blood Agar with 5% Rabbit Blood can be used in qualitative procedures for the cultivation of Haemophilus species.

  • Rabbit blood in the medium does not contain V factor -inactivating enzymes and supplies X and V factors, one or both of which are required for the growth of Haemophilus Rabbit blood also allows for differentiation of hemolytic and nonhemolytic species of Haemophilus.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
DescriptionBlood Agar (TSA w/5% Rabbit Blood)
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach