Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Isolate and cultivate fastidious aerobic as well as anaerobic microorganisms with Thermo Scientific™ Remel™ Brucella Agar with 5% Sheep Blood. It is prepared according to the formulation of the American Public Health Association for Albimi Broth1,2 and recommended for isolation obligate and facultative anaerobes from clinical specimens3,4.
Description | Brucella Agar w/5% Sheep Blood |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01252 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Casein and meat peptones in the medium supply nitrogen, amino acids, and peptides necessary for bacterial growth.Dextrose is a ready source of energy, Yeast extract provides B-complex vitamins and enhances growth. Sodium chloride us a sorce of essential electrolytes and maintains osmotic equilibrium. Sodium bisulfite is a reducing agent. Sheep blood provides growth factors required by certain anaerobic organisms and allows for demonstration of hemolysis by aerobic and anerobic bacteria. Agar is a solidifying agent.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.