Remel™ Charcoal Blood Agar w/cephalexin (Regan-Lowe Medium)
Thermo Scientific™
Remel™ Charcoal Blood Agar w/cephalexin (Regan-Lowe Medium)
Use for selective isolation of Bordetella pertussis and Bordetella parapertussis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatMedia TypeIncludes
R01298MonoplateCharcoal Blood AgarCephalexin
R100551 oz. BottleCharcoal Blood AgarCephalexin
Catalog number R01298
Price (USD)
-
Format:
Monoplate
Media Type:
Charcoal Blood Agar
Includes:
Cephalexin
Thermo Scientific™ Remel Charcoal Blood Agar w/cephalexin (Regan-Lowe Medium) is used for selective isolation of Bordetella pertussis and Bordetella parapertussis.
Specifications
FormatMonoplate
IncludesCephalexin
Media TypeCharcoal Blood Agar
Quantity10/Pk.
Product TypeAgar
Unit SizeEach