Inquire about OEM or Commercial Supply version of this product here.
dUTP Solution (100 mM)
Thermo Scientific™

dUTP Solution (100 mM)

Thermo Scientific™ dUTP (2'-deoxyuridine 5'-triphosphate) is highly stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.
Have Questions?
Catalog NumberQuantity
R01330.25 mL
Catalog number R0133
Price (USD)
54.90
Each
Quantity:
0.25 mL
Request bulk or custom format
Price (USD)
54.90
Each

Thermo Scientific™ dUTP (2'-deoxyuridine 5'-triphosphate) is highly stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. The nucleotide has greater than 99% purity and is free of nuclease activities and human and E. coli DNA. It is designed for many different molecular biology applications.

Features

  • Greater than 99% purity confirmed by HPLC
  • Free of human and E. coli DNA
  • Highly stable

Applications

  • Long range PCR (40 kb)
  • cDNA synthesis and RT-PCR
  • Real-time PCR
  • Standard PCR
  • High fidelity PCR
  • Primer extension.

Note: dUTP is not suitable for PCR with proofreading enzymes or enzyme mixes containing proofreading enzymes.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
PolymeraseDNA Polymerase
Purity or Quality Grade>99%
Quantity0.25 mL
Shipping ConditionDry Ice
Concentration100 mM
For Use With (Application)Standard PCR, High-fidelity PCR, RT-PCR, Real Time PCR (qPCR)
FormLiquid
Label or DyeUnlabeled
PCR Method2-step RT-qPCR, Standard PCR
Unit SizeEach
Contents & Storage
dUTP Solution, 100 mM (0.25 mL)

Store at –20°C.