Thermo Scientific™

Remel™ Buffered CYE Differential Agar w/BTB, BCP

Catalog number: R01343
Thermo Scientific™

Remel™ Buffered CYE Differential Agar w/BTB, BCP

Catalog number: R01343

Thermo Scientific™ Remel Buffered CYE Differential Agar w/BTB, BCP (BCYE w/Dyes) is used for qualitative procedures to isolate Legionella species.

 
Catalog Number
R01343
Unit Size
Each
dimFormat
Price (USD)
Full specifications
DescriptionBuffered CYE Differential Agar w/BTB, BCP
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizedimFormatPrice (USD)
R01343Full specifications
EachRequest A Quote
DescriptionBuffered CYE Differential Agar w/BTB, BCP
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Showing 1 of 1
  • Contains dyes to differentiate Tatlockia spp.from Legionella pneumophila