Thermo Scientific™

Cornmeal Agar with Polysorbate 80 with or without Trypan Blue

Catalog number: R01354
Thermo Scientific™

Cornmeal Agar with Polysorbate 80 with or without Trypan Blue

Catalog number: R01354

Cultivate fungi and stimulate chlamydospore production by Candida albicans with Thermo Scientific™ Remel Cornmeal Agar with Polysorbate 80 general purpose medium, with or without Trypan Blue. Polack and Benham demonstrated the use of cornmeal agar for studying differential mycelial characteristics of Candida species1,2. Adding polysorbate 80 to cornmeal agar stimulates chlamydospore formation by Candida albicans3-4.

 
Catalog Number
R01354
Unit Size
Each
Format
Monoplate Deep Pour
Price (USD)
Full specifications
DescriptionCornmeal Agar w/Tween™ 80
FormatMonoplate Deep Pour
Product TypeAgar
TypeAgar
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
12mm x 85mm Monoplate
Price (USD)
Full specifications
DescriptionCornmeal Agar w/Tween™ 80
Format12mm x 85mm Monoplate
Product TypeAgar
TypeAgar
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 150mm Pour Tube
Price (USD)
Full specifications
DescriptionCornmeal Agar w/Tween™ 80
Format20mm x 150mm Pour Tube
Product TypeAgar
TypeAgar
Quantity20/Pk.
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeFormatPrice (USD)
R01354Full specifications
EachMonoplate Deep PourRequest A Quote
DescriptionCornmeal Agar w/Tween™ 80
FormatMonoplate Deep Pour
Product TypeAgar
TypeAgar
Quantity10/Pk.
Unit SizeEach
R01328Full specifications
Each12mm x 85mm MonoplateRequest A Quote
DescriptionCornmeal Agar w/Tween™ 80
Format12mm x 85mm Monoplate
Product TypeAgar
TypeAgar
Quantity10/Pk.
Unit SizeEach
R09290Full specifications
Each20mm x 150mm Pour TubeRequest A Quote
DescriptionCornmeal Agar w/Tween™ 80
Format20mm x 150mm Pour Tube
Product TypeAgar
TypeAgar
Quantity20/Pk.
Unit SizeEach
Showing 3 of 3

Cornmeal Agar with Polysorbate 80 with or without trypan blue is a solid medium recommended for use in qualitative procedures as a general-purpose media for cultivating fungi and stimulating chlamydospore production by Candida albicans.

  • General-purpose media: cultivate fungi and stimulate chlamydospore formation by Candida albicans.
  • Easy-to-read: Trypan blue provides a contrasting background for the optimal visualization of chlamydospores.
  • Ready-to-use: the convenience of prepared media.

Cornmeal agar consists of an infusion of cornmeal and agar, which provides the essential nutrients needed to support the growth of fungi and the production of chlamydospores by Candida albicans. The agar is a solidifying agent. Polysorbate 80 reduces surface tension and enhances the development of chlamydospores. Trypan blue is an optimal additive that provides a contrasting background, thereby permitting optimal visualization of chlamydospores.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Figures