Mueller Hinton Agar
Thermo Scientific™

Mueller Hinton Agar

Perform effortless Antimicrobial Susceptibility Testing (AST) using this medium which may be used in internationally recognized standard procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R01624100/Cs.15 x 100mm Plate
R0162010/Pk.15 x 100mm Plate
R0405010/Pk.15 x 150mm Plate
R0405240/Pk.15 x 150mm Plate
Catalog number R01624
Price (USD)
-
Quantity:
100/Cs.
Format:
15 x 100mm Plate
Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Mueller Hinton Agar, a medium designed and manufactured according to the European standard, EUCAST, and the American standard, CLSI.
The addition of starch to Mueller Hinton Agar ensures that toxic factors found during growth will be absorbed, and its presence is often essential to establish growth from very small inocula. Conforms to EUCAST, CLSI and DIN.

Our products are manufactured and distributed across the world through a vast network of company owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery We recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMueller Hinton Agar
Format15 x 100mm Plate
Quantity100/Cs.
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach