Search Thermo Fisher Scientific
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons with Gentamicin is used for the selective isolation of yeast and fungi.
Description | Sabouraud Dextrose Agar, Emmons w/Gentamicin |
Format | Monoplate Deep Pour |
Product Type | Agar |
Type | Agar |
Quantity | 20/Pk. |
Unit Size | Each |
Description | Sabouraud Dextrose Agar, Emmons w/Gentamicin |
Format | 20mm x 113mm Tube |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Format | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01776 | Full specifications | Each | Monoplate Deep Pour | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R08738 | Full specifications | Each | 20mm x 113mm Tube | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|