TBS Buffer

Catalog number: R017R.0000

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tris Buffered Saline (TBS) Buffer is intended for use with PanPath products. TBS is supplied as a lyophilized powder, and when dissolved in one liter distilled or deionised water, it becomes 0.05M TBS, pH 8.0 (0.238M NaCl; 0.0027M KCl) with 0.05% Tween 20.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Powder
Product Line: REMBRANDT®
Product Size: 1 pack
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols