SXT Blood Agar
Thermo Scientific™
SXT Blood Agar
Selectively isolate beta-hemolytic Group A Streptococci from throat cultures and upper respiratory specimens using SXT Blood Agar.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R0186010/Pk.
R01862100/Pk.
Catalog number R01860
Price (USD)
-
Quantity:
10/Pk.
Selectively isolate beta-hemolytic group A streptococci from throat cultures and upper respiratory specimens using Thermo Scientific™ Remel™ SXT Blood Agar. Pacifico at al. demonstrated the isolation of β-hemolyic streptococci is improved with the use of a selective medium which inhibits oropharyngeal commensal microbial flora1. In 1977, Gunn et al. introduced a selective medium containing sulfamethoxazole and trimethoprim in a tryptic soy agar base supplemented with 5% sheep blood2. In 1979, Kurzynski et al. proved that streptococci were more frequently isolated on SXT sheep blood agar3.
Clinical assessment alone is unreliable in distinguishing streptococcal and non-streptococcol pharyngitis4. Serious medical complications may follow pharyngitis caused by group A streptococci, making bacterial examination of the throat an indispensable diagnostic tool.
Use of SXT sheep blood agar allows enhanced recovery of Lancefield groups A and B streptococci from throat cultures while inhibiting the growth of commensal microbial flora.
  • Ready-to-use – The convenience of prepared media
  • Selective – Use of sulfamethoxazole plus trimethoprim (SXT) supresses growth of streptococcal groups C, F, and G and Streptococci viridans, most Enterobacteriaceae, Neisseria, and some Pseudomonas
This medium contains casein and soy peptones that provide nutritious source of nitrogen, amino acids, and peptides. Sheep blood provides growth factors and allows detection of hemolytic reactions. Sodium chloride maintains osmotic equilibrium. SXT (sulfamethoxazole, trimethoprim) act as selective agents to supresse most commensal microbial flora found in the oropharynx, including streptococcal groups C, F, and G and Streptococci viridans, most Enterobacteriaceae, Neisseria, and some Pseudomonas spp.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. L., A. Ranucci, G. Ravagnan, and C. Chiesa. 1995. J. Clin. Microbiol. 33:2480-2482.
  2. Gunn, B.A., D.K. Ohashi, C.A. Gaydos, and E.S. Holt. 1977. J. Clin. Microbiol. 5:650-655.
  3. Kurzynski, T.A. and C.K. Meise. 1979. J. Clin. Microbiol. 9:189-193.
  4. Murray, P.R., E.J. Baron., J.H. Jorgensen, M.L. Landry, and M.A. Pfaller. 2007. Manual of Clinical Microbiology. 9th ed. ASM Press, Washington, D.C.
Specifications
DescriptionSXT Blood Agar w/ sulfamethoxazole, trimethoprim
Format12mm x 85mm Monoplate
Quantity10/Pk.
Product TypeAgar Plate
Unit SizeEach