| R01917 | Full specifications | Each | | 100/Pk. | 100/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, SMWW. | Description | Tryptic Soy Agar | Format | Monoplate | Product Type | Agar | Packaging Type | 100/Pk. | Quantity | 100/Pk. | Unit Size | Each |
|
| R01920 | Full specifications | Each | | 10/Pk. | 10/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | Monoplate | Product Type | Agar | Packaging Type | 10/Pk. | Quantity | 10/Pk. | Unit Size | Each |
|
| R04080 | Full specifications | Each | | 10/Pk. | 10/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 15mm x 150mm Plate | Product Type | Agar | Packaging Type | 10/Pk. | Quantity | 10/Pk. | Unit Size | Each |
|
| R064860 | Full specifications | Each | | 100/Pk. | 100/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 15mm x 103mm Tube | Product Type | Agar | Packaging Type | 100/Pk. | Quantity | 100/Pk. | Unit Size | Each |
|
| R064862 | Full specifications | Each | | 20/Pk. | 20/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 15mm x 103mm Tube | Product Type | Agar | Packaging Type | 20/Pk. | Quantity | 20/Pk. | Unit Size | Each |
|
| R08930 | Full specifications | Each | | 100/Pk. | 100/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 20mm x 113mm Tube | Product Type | Agar | Packaging Type | 100/Pk. | Quantity | 100/Pk. | Unit Size | Each |
|
| R08932 | Full specifications | Each | | 20/Pk. | 20/Pk. | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 20mm x 113mm Tube | Product Type | Agar | Packaging Type | 20/Pk. | Quantity | 20/Pk. | Unit Size | Each |
|
| R08934 | Full specifications | Each | | 100/Pk. | 100 x 10 mL | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 20mm x 113mm Pour Tube | Product Type | Agar | Packaging Type | 100/Pk. | Quantity | 100 x 10 mL | Unit Size | Each |
|
| R09938 | Full specifications | Each | | 100/Pk. | 100 x 22 mL | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 20mm x 150mm Pour Tube | Product Type | Agar | Packaging Type | 100/Pk. | Quantity | 100 x 22 mL | Unit Size | Each |
|
| R09940 | Full specifications | Each | | 20/Pk. | 20 x 22 mL | Request A Quote |
Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | Description | Tryptic Soy Agar | Format | 20mm x 150mm Pour Tube | Product Type | Agar | Packaging Type | 20/Pk. | Quantity | 20 x 22 mL | Unit Size | Each |
|