Thermo Scientific™

Remel™ Tryptic Soy Agar

Catalog number: R01921
Thermo Scientific™

Remel™ Tryptic Soy Agar

Catalog number: R01921

Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

 
Catalog Number
R01921
Unit Size
Each
Format
Spacesaver Plate
Packaging Type
15/Pk.
Quantity
15/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
FormatSpacesaver Plate
Product TypeAgar
TypeAgar
Packaging Type15/Pk.
Quantity15/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
Monoplate
Packaging Type
100/Pk.
Quantity
100/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, SMWW.
DescriptionTryptic Soy Agar
FormatMonoplate
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
Monoplate
Packaging Type
10/Pk.
Quantity
10/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
FormatMonoplate
Product TypeAgar
TypeAgar
Packaging Type10/Pk.
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 150mm Plate
Packaging Type
10/Pk.
Quantity
10/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format15mm x 150mm Plate
Product TypeAgar
TypeAgar
Packaging Type10/Pk.
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Packaging Type
100/Pk.
Quantity
100/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Packaging Type
20/Pk.
Quantity
20/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Packaging Type20/Pk.
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Packaging Type
100/Pk.
Quantity
100/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Packaging Type
20/Pk.
Quantity
20/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Packaging Type20/Pk.
Quantity20/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Pour Tube
Packaging Type
100/Pk.
Quantity
100 x 10 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 113mm Pour Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100 x 10 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 150mm Pour Tube
Packaging Type
100/Pk.
Quantity
100 x 22 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 150mm Pour Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100 x 22 mL
Unit SizeEach
Showing 10 of 14Show more
Catalog NumberSpecificationsUnit SizeFormatPackaging TypeQuantityPrice (USD)
R01921Full specifications
EachSpacesaver Plate15/Pk.15/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
FormatSpacesaver Plate
Product TypeAgar
TypeAgar
Packaging Type15/Pk.
Quantity15/Pk.
Unit SizeEach
R01917Full specifications
EachMonoplate100/Pk.100/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, SMWW.
DescriptionTryptic Soy Agar
FormatMonoplate
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100/Pk.
Unit SizeEach
R01920Full specifications
EachMonoplate10/Pk.10/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
FormatMonoplate
Product TypeAgar
TypeAgar
Packaging Type10/Pk.
Quantity10/Pk.
Unit SizeEach
R04080Full specifications
Each15mm x 150mm Plate10/Pk.10/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format15mm x 150mm Plate
Product TypeAgar
TypeAgar
Packaging Type10/Pk.
Quantity10/Pk.
Unit SizeEach
R064860Full specifications
Each15mm x 103mm Tube100/Pk.100/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100/Pk.
Unit SizeEach
R064862Full specifications
Each15mm x 103mm Tube20/Pk.20/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Packaging Type20/Pk.
Quantity20/Pk.
Unit SizeEach
R08930Full specifications
Each20mm x 113mm Tube100/Pk.100/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100/Pk.
Unit SizeEach
R08932Full specifications
Each20mm x 113mm Tube20/Pk.20/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 113mm Tube
Product TypeAgar
TypeAgar
Packaging Type20/Pk.
Quantity20/Pk.
Unit SizeEach
R08934Full specifications
Each20mm x 113mm Pour Tube100/Pk.100 x 10 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 113mm Pour Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100 x 10 mL
Unit SizeEach
R09938Full specifications
Each20mm x 150mm Pour Tube100/Pk.100 x 22 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
DescriptionTryptic Soy Agar
Format20mm x 150mm Pour Tube
Product TypeAgar
TypeAgar
Packaging Type100/Pk.
Quantity100 x 22 mL
Unit SizeEach
Showing 10 of 14Show more

Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

REF R110092, REF R112700, and REF R112703 not intended for IVD use.

Figures