Thermo Scientific™

V Agar Monoplate

Catalog number: R01950
Thermo Scientific™

V Agar Monoplate

Catalog number: R01950

Isolate and identify Gardnerella vaginalis with Thermo Scientific™ Remel™ V Agar prepared plates, a medium containing a special blood agar base with 5% human blood, designed to enhance the growth of fastidious organisms.

 
Catalog Number
R01950
Unit Size
Each
Quantity
10/Pk.
Price (USD)
Full specifications
DescriptionV Agar
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
100/Pk.
Price (USD)
Full specifications
DescriptionV Agar
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity100/Pk.
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R01950Full specifications
Each10/Pk.Request A Quote
DescriptionV Agar
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
R01960Full specifications
Each100/Pk.Request A Quote
DescriptionV Agar
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity100/Pk.
Unit SizeEach
Showing 2 of 2

Use the solid media V Agar Monoplate to isolate and identify Gardnerella vaginalis.

  • Promotes: V Agar contains the essential elements for the growth of Gardnerella vaginalis.
  • Easy to read: The transparent agar allows differentiation of G. vaginalis colonies from those of commensal microbial flora (e.g., streptococci, yeasts, lactobacilli, etc.).
  • Selective: Colistin, nalidixic acid and nystatin inhibit most gram-negative bacilli, yeast and diptheroids.
  • Ready-to-use: Convenience of prepared media.

Casein and meat peptones supply nitrogenous compounds, carbon, sulphur and trace elements necessary for the growth of G. vaginalis. Cornstarch is an energy source. Human blood is incorporated into the agar to allow for presumptive identification of G. vaginalis, which produces diffuse beta-hemolysis on V agar and Agar Selective. Colistin, nalidixic acid and nystatin are selective agents, which inhibit most gram-negative bacilli, yeast and diptheroids.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text