Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
Description | MacConkey Agar//MacConkey Agar w/Sorbitol |
Product Type | Agar |
Quantity | 10/Pk. |
Type | Agar |
Format | Biplate |
Unit Size | Each |
Description | HE Agar; XLD Agar |
Format | Biplate |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | MacConkey Agar//PEA Sheep Blood Agar |
Product Type | Agar |
Quantity | 10/Pk. |
Type | Agar |
Format | Biplate |
Unit Size | Each |
Description | Middlebrook 7H11 Agar//Mitchison 7H11 Selective |
Format | Biplate |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | Middlebrook 7H10/Mitchison 7H11 10/PK |
Format | Biplate |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | V Agar; Starch Agar w/Brom Cresol Purple; Biplate; 10/Pk. |
Format | Biplate |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Description | QC-Slide™ AFB Stain Control; 10/Pk. |
Product Type | Agar |
Quantity | 10/Pk. |
Type | Agar |
Format | Biplate |
Unit Size | Each |
Description | SXT Blood Agar and SXT Blood Agar (TSA w/5% Sheep Blood) |
Product Type | Agar |
Quantity | 10/Pk. |
Type | Agar |
Format | Biplate |
Unit Size | Each |
Description | Tergitol 7 Agar with TTC; Blood Agar |
Format | Biplate |
Product Type | Agar |
Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R02095 | Each | MacConkey Agar//MacConkey Agar w/Sorbitol | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02086 | Each | HE Agar; XLD Agar | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02096 | Each | MacConkey Agar//PEA Sheep Blood Agar | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02112 | Each | Middlebrook 7H11 Agar//Mitchison 7H11 Selective | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02118 | Each | Middlebrook 7H10/Mitchison 7H11 10/PK | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02148 | Each | V Agar; Starch Agar w/Brom Cresol Purple; Biplate; 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02140 | Each | QC-Slide™ AFB Stain Control; 10/Pk. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02142 | Each | SXT Blood Agar and SXT Blood Agar (TSA w/5% Sheep Blood) | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R02144 | Each | Tergitol 7 Agar with TTC; Blood Agar | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.