Haemophilus ID Quad Medium
Thermo Scientific™

Haemophilus ID Quad Medium

Differentiate and determine Haemophilus species efficiently using a single plate with four separate media formulations.
Have Questions?
Catalog NumberdimFormatQuantityFill VolumeManufacturing Origin
R0246010/Pk.5 mLUnited States
Catalog number R02460
Price (USD)
-
Quantity:
10/Pk.
Fill Volume:
5 mL
Manufacturing Origin:
United States
Speciate the genus Haemophilus using Thermo Scientific™ Haemophilus ID Quad, a medium containing three separate growth requirements and horse blood for the production of hemolysis, all on a single prepared media plate.

Read more results from a single sample and produce more diagnostic details while reducing storage capacity issues in the incubator with a quad-plate that includes four separate media formulations.

  • One quadrant of the plate contains X factor
  • One quadrant contains V factor
  • One quadrant contains XV factors
  • One quadrant contains XV factors with horse blood

Haemophilus are small, pleomorphic, gram-negative coccobacilli. They are nonmotile, non-sporeforming, fastidious, facultative anaerobes. Almost all species require preformed growth factors present in the blood, particularly the X factor (hemin) and/or V factor (NAD) for growth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
ColorClear agar light to med amber/ clear agar light amber/ clear agar light to med amber/ Opaque red
DescriptionHaemophilus ID Quad Medium
Fill Volume5 mL
Format85mm Quadplate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life6 Weeks
Manufacturing OriginUnited States
Product TypeAgar Plate
Unit SizeEach
Contents & Storage
2°C to 8°C