Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Clear agar |
Description | Synergy Quad |
Format | 85mm Quadplate |
Manufacturing Origin | United States |
Product Type | Agar |
Fill Volume | 5 mL |
Packaging Type | Flow wrapped - single layer |
Quantity | 10/Pk. |
Shelf Life | 8 Weeks |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Quantity | Fill Volume | Manufacturing Origin | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R02474 | Each | 10/Pk. | 5 mL | United States | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|