dm5CTP Solution (10 mM)

Catalog number: R0431

Thermo Scientific™  Related applications: Nucleic Acid Labeling & Oligo Synthesis | PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific dm5CTP (2'-deoxy-5'-methylcytidine 5'-triphosphate) modified nucleotide is used in synthesis of methylated DNA. It supplied as 10 mM aqueous solution titrated to pH 7.0 with NaOH.

dm5CTP can be enzymatically incorporated into DNA with Taq DNA polymerase, Klenow Fragment.

Applications

• Synthesis of methylated DNA
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration (molarity): 10 mM
Label or Dye: Methylated
Labeling Method: Direct Labeling
Product Size: 0.1 mL

Documents

Manuals & protocols

Recommended products