TheraPure™ GMP Nucleotides
Thermo Scientific™

TheraPure™ GMP Nucleotides

TheraPure GMP nucleotides help accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo de productoConcentraciónCantidadFormato
R044SKB002ATP, solución de sodio100 mM17 mLSuministro comercial
R00014SKB002ATP, solución Tris200 mM5 mLSuministro comercial
R00014SKB005ATP, solución Tris200 mM50 mLSuministro comercial
R00014SKB004ATP, solución Tris200 mM500 mLSuministro comercial
R044SKB001ATP, solución de sodio100 mM3,5 mLSuministro comercial
R0441SKB003ATP, solución de sodio100 mM50 mLSuministro comercial
R0441SKB009ATP, solución de sodio100 mM500 mLSuministro comercial
R00015SKB002CTP, solución Tris200 mM5 mLSuministro comercial
R00015SKB005CTP, solución Tris200 mM50 mLSuministro comercial
R00015SKB004CTP, solución Tris200 mM500 mLSuministro comercial
R045SKB001CTP, solución de sodio100 mM3,5 mLSuministro comercial
R045SKB002CTP, solución de sodio100 mM17 mLSuministro comercial
R0451SKB003CTP, solución de sodio100 mM50 mLSuministro comercial
R0451SKB009CTP, solución de sodio100 mM500 mLSuministro comercial
R00016SKB002GTP, solución Tris200 mM5 mLSuministro comercial
R00016SKB005GTP, solución Tris200 mM50 mLSuministro comercial
R00016SKB004GTP, solución Tris200 mM500 mLSuministro comercial
R046SKB001GTP, solución de sodio100 mM3,5 mLSuministro comercial
R046SKB002GTP, solución de sodio100 mM20,5 mLSuministro comercial
R0461SKB003GTP, solución de sodio100 mM50 mLSuministro comercial
R0461SKB009GTP, solución de sodio100 mM500 mLSuministro comercial
R00017SKB002UTP, solución Tris200 mM5 mLSuministro comercial
R00017SKB005UTP, solución Tris200 mM50 mLSuministro comercial
R00017SKB004UTP, solución Tris200 mM500 mLSuministro comercial
R047SKB001UTP, solución de sodio100 mM3,5 mLSuministro comercial
R0471SKB006UTP, solución de sodio100 mM20 mLSuministro comercial
R0471SKB002UTP, solución de sodio100 mM100 mLSuministro comercial
R0471SKB003UTP, solución de sodio100 mM500 mLSuministro comercial
R00014SKB002CATP, solución Tris200 mM5 mLCompra instantánea
R044SKB001CATP, solución de sodio100 mM3,5 mLCompra instantánea
R00015SKB002CCTP, solución Tris200 mM5 mLCompra instantánea
R045SKB001CCTP, solución de sodio100 mM3,5 mLCompra instantánea
R00016SKB002CGTP, solución Tris200 mM5 mLCompra instantánea
R046SKB001CGTP, solución de sodio100 mM3,5 mLCompra instantánea
R00017SKB002CUTP, solución Tris200 mM5 mLCompra instantánea
R047SKB001CUTP, solución de sodio100 mM3,5 mLCompra instantánea
Número de catálogo R044SKB002
Precio (EUR)
-
Tipo de producto:
ATP, solución de sodio
Concentración:
100 mM
Cantidad:
17 mL
Formato:
Suministro comercial
Thermo Scientific TheraPure GMP nucleoside triphosphates (NTPs) are manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. They are suitable as core building blocks for mRNA synthesis in the development and manufacturing of mRNA therapeutics. Nucleotides ATP, CTP, GTP, and UTP are available and are supplied in sodium- or Tris-titrated solutions.

Thermo Scientific TheraPure GMP* nucleoside triphosphates (NTPs) are manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. They are suitable as core building blocks for mRNA synthesis in the development and manufacturing of mRNA therapeutics. Nucleotides ATP, CTP, GTP, and UTP are available and are supplied in sodium- or Tris-titrated solutions.

Designed for process development and manufacturing

TheraPure GMP nucleotides help accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization by providing:

  • Quality—meet critical product quality needs to accelerate therapeutic development and manufacture
  • Minimal risk—reduce risks in developing a therapeutic by utilizing proven products
  • Technical partnership—partner with expert product and technical support
  • Secure and consistent supply—count on a secure and stable supply of product at a global scale

Supported by unique quality system

TheraPure GMP nucleotides are supported by a comprehensive quality system and documentation, including:

  • Manufactured to relevant ICH Q7 GMP guidelines
  • Animal-origin–free (AOF) manufacturing processes, materials, and facilities
  • Validated manufacturing processes and analytical test methods
  • Comprehensive impurity profiles
  • Verified compendial assays where applicable
  • Product stability data
  • Tested by analytical methods following ICH Q2 guidelines whenever possible
  • Drug master file (DMF)

Two available formats

These two formats are offered to accommodate different mRNA manufacturing processes:

  • 100 mM aqueous solution with Na+ titrated to pH 7.3–7.5 with NaOH
  • 200 mM aqueous solution with Tris counterion titrated to pH 7.3–7.7 with Tris base

Modified nucleotides

TheraPure GMP N1-methylpseudo-UTP is also available.

*TheraPure GMP refers to the quality level of the raw, ancillary, or starting materials to be used for further manufacturing. TheraPure GMP products are manufactured in facilities with ISO 9001–certified quality management systems that operate in accordance with relevant good manufacturing practice (GMP) principles, as outlined in ICH Q7 or equivalent guidance documents or standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
ColorTransparent
Para utilizar con (aplicación)IVT Transcription, RNA Synthesis, mRNA Synthesis
FormatoSuministro comercial
Etiqueta o tinteSin marcar
Peso molecular507.18 g/mol
Tipo de productoATP, solución de sodio
Pureza≥ 99%
Cantidad17 mL
Duración de almacenamientoUp to 36 months after manufacturing when stored at –20°C
Concentración100 mM
Línea de productosTheraPure™ GMP
pH7.3 to 7.5
Unit SizeEach