TheraPure™ Nucleotides
Thermo Scientific™

TheraPure™ Nucleotides

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호제품 유형농도
R045T1CTP Solution, Sodium Solution100 mM
R044T1ATP Solution, Sodium Solution100 mM
R00044T1ATP Solution, Tris Solution200 mM
R00045T1CTP Solution, Tris Solution200 mM
R046T1GTP Solution, Sodium Solution100 mM
R00046T1GTP Solution, Tris Solution200 mM
R047T1UTP Solution, Sodium Solution100 mM
R00047T1UTP Solution, Tris Solution200 mM
R0491T1N1-Methylpseudo-UTP, Sodium Solution100 mM
카탈로그 번호 R045T1
제품 가격(KRW)
79,000
온라인 행사
Ends: 31-Mar-2026
92,000
할인액 13,000 (14%)
Each
제품 유형:
CTP Solution, Sodium Solution
농도:
100 mM
제품 가격(KRW)
79,000
온라인 행사
Ends: 31-Mar-2026
92,000
할인액 13,000 (14%)
Each
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement
For Research Use Only.
사양
색상Colorless
용도(애플리케이션)In vitro Transcription
제품 유형CTP Solution, Sodium Solution
수량0.2 mL
배송 조건Dry Ice
농도100 mM
제품라인TheraPure
Unit SizeEach
구성 및 보관
Store at -20°C.