Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Differentiate anaerobic bacteria using Thermo Scientific™ Remel PRAS (Pre-Reduced Anaerobic Sterilized) PY Media.
Description | PRAS PY Glucose |
Format | 15 x 103mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 10 mL |
Unit Size | Each |
Description | PRAS PY Base Control |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Description | PRAS PY Bile |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Description | PRAS PY Glucose |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Description | PRAS PY Inositol |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Description | PRAS PY Bile |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 100/Pk. |
Quantity | 100 x 3 mL |
Unit Size | Each |
Description | PRAS PY Arabinose |
Format | 16mm x 125mm Tube, Rubber Stopper |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Description | P.R.A.S. PY Esculin |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 100/Pk. |
Quantity | 100 x 3 mL |
Unit Size | Each |
Description | P.R.A.S. PY Glucose Agar ; Format: 16 x 125mm slant; pack of 20 |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Description | P.R.A.S. PY Galactose |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Packaging Type | 20/Pk. |
Quantity | 20 x 3 mL |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | dimFormat | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R05194 | Each | PRAS PY Glucose | 20 x 10 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05161 | Each | PRAS PY Base Control | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05175 | Each | PRAS PY Bile | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05191 | Each | PRAS PY Glucose | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05205 | Each | PRAS PY Inositol | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05176 | Each | PRAS PY Bile | 100 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05171 | Each | PRAS PY Arabinose | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05181 | Each | P.R.A.S. PY Esculin | 100 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05192 | Each | P.R.A.S. PY Glucose Agar ; Format: 16 x 125mm slant; pack of 20 | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R05189 | Each | P.R.A.S. PY Galactose | 20 x 3 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Differentiate anaerobic bacteria using Thermo Scientific™ Remel PRAS (Pre-Reduced Anaerobic Sterilized) PY Media.