TurboFect™ Transfection Reagent
Thermo Scientific™

TurboFect™ Transfection Reagent

Thermo Scientific TurboFect Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expressionRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R05311 mL
R0533200 μL
R05345 x 200 μL
R05325 x 1 mL
Catalog number R0531
Price (USD)
436.00
Each
Quantity:
1 mL
Price (USD)
436.00
Each
Thermo Scientific TurboFect Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.

Features of TurboFect Transfection Reagent:

• High transfection efficiency of a wide variety of cell types
• No optimization required for most cell lines
• Excellent transfection efficiency in the presence or absence of serum
• Minimal cytotoxicity
• Ready-to-use—no need to reconstitute, dilute or manipulate

Applications of TurboFect Transfection Reagent :
• Stable and transient transfection or co-transfection of DNA or RNA
• Experiments with adherent or suspension cells, differentiated or undifferentiated cells
• Gene expression studies and assays involving GFP or luciferase reporters

TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. TurboFect Reagent interacts with DNA to form small, stable, highly diffusible complexes which are readily endocytosed. The reagent creates a proton-sponge effect that buffers endosomal pH by provoking massive proton accumulation and passive chloride influx. As a result, rapid osmotic swelling of the endocytosed DNA-complex causes it to rupture in the cytosol, allowing translocation of DNA to the nucleus. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationChemically Defined
Compatible CellsPrimary Cell, Endothelial Cell, Epithelial Cell, Myeloma Cell, Cell Lines
For Use With (Application)Transfection
Format6-Well Plate, 12-Well Plate, 24-Well Plate, 48-Well Plate, 96-Well Plate, Flasks
Product LineTurboFect
Quantity1 mL
Transfection TechniqueChemical Transfection
TypeTransfection Reagent
Unit SizeEach

Frequently asked questions (FAQs)

What is the stability of TurboFECT after being frozen?

TurboFect is a sterile solution of a proprietary cationic polymer in water. Its performance is not affected by a few freeze-thaw cycles. If TurboFECT arrives frozen or is accidentally stored at -20 degrees C, simply thaw and mix it thoroughly before use. For long-term storage, keep TurboFECT at 4 degrees C as recommended.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.