Thermo Scientific™
Remel™ Andrade's Broth
Related applications:
Clinical Microbiology
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.
Back to top
Description
Thermo Scientific™ Remel Andrades Broth is used for the determination of fermentation reactions of microorganisms, particularly Enterobacteriaceae.