Thermo Scientific™
Brain Heart Infusion (BHI) Agar
Related applications:
Clinical Microbiology
Isolate and cultivate a wide variety of microorganisms including pathogenic fungi with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar.  Due to the addition of 5-10% sheep blood, BHI agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media. BHI Agar is recommended by the Association of Official Analytical Chemists (AOAC) and the American Public Health Association (APHA) 1, 2
Back to top
Use Heart Infusion Agar for the cultivation of a wide variety of microorganisms including pathogenic fungi.
  • Recommended by AOAC and APHA.
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
REF R112025 not intended for IVD use.