Thermo Scientific™

Remel™ BHI Broth (Brain Heart Infusion Broth)

Catalog number: R060252
Thermo Scientific™

Remel™ BHI Broth (Brain Heart Infusion Broth)

Catalog number: R060252

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

 
Catalog Number
R060252
Unit Size
Each
Format
15mm x 103mm Tube - Filtered
Quantity
20 x 0.5 mL
Price (USD)
Full specifications
Format15mm x 103mm Tube - Filtered
TypeBroth
DescriptionBHI Broth
Product TypeBroth
Quantity20 x 0.5 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube - Filtered
Quantity
100 x 0.5 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube - Filtered
Product TypeBroth
TypeBroth
Quantity100 x 0.5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
100 x 2 mL
Price (USD)
Full specifications
Format15mm x 103mm Tube
TypeBroth
DescriptionBHI Broth
Product TypeBroth
Quantity100 x 2 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
20 x 2 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionMIC Susceptibility System, BHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity20 x 2 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
100 x 4 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity100 x 4 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
20 x 4 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity20 x 4 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
100 x 5 mL
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity100 x 5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Format
15mm x 103mm Tube
Quantity
20 x 5 mL
Price (USD)
Full specifications
Format15mm x 103mm Tube
TypeBroth
DescriptionBHI Broth
Product TypeBroth
Quantity20 x 5 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Quantity
100 x 10 mL
Price (USD)
Full specifications
Format20mm x 113mm Tube
TypeAgar
DescriptionBHI Broth
Product TypeAgar
Quantity100 x 10 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
Catalog Number
Unit Size
Each
Format
20mm x 113mm Tube
Quantity
20 x 10 mL
Price (USD)
Full specifications
Format20mm x 113mm Tube
TypeAgar
DescriptionBHI Broth
Product TypeAgar
Quantity20 x 10 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeFormatQuantityPrice (USD)
R060252Full specifications
Each15mm x 103mm Tube - Filtered20 x 0.5 mLRequest A Quote
Format15mm x 103mm Tube - Filtered
TypeBroth
DescriptionBHI Broth
Product TypeBroth
Quantity20 x 0.5 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
R060250Full specifications
Each15mm x 103mm Tube - Filtered100 x 0.5 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube - Filtered
Product TypeBroth
TypeBroth
Quantity100 x 0.5 mL
Unit SizeEach
R060258Full specifications
Each15mm x 103mm Tube100 x 2 mLRequest A Quote
Format15mm x 103mm Tube
TypeBroth
DescriptionBHI Broth
Product TypeBroth
Quantity100 x 2 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
R060260Full specifications
Each15mm x 103mm Tube20 x 2 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionMIC Susceptibility System, BHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity20 x 2 mL
Unit SizeEach
R060264Full specifications
Each15mm x 103mm Tube100 x 4 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity100 x 4 mL
Unit SizeEach
R060266Full specifications
Each15mm x 103mm Tube20 x 4 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity20 x 4 mL
Unit SizeEach
R060270Full specifications
Each15mm x 103mm Tube100 x 5 mLRequest A Quote
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Product TypeBroth
TypeBroth
Quantity100 x 5 mL
Unit SizeEach
R060272Full specifications
Each15mm x 103mm Tube20 x 5 mLRequest A Quote
Format15mm x 103mm Tube
TypeBroth
DescriptionBHI Broth
Product TypeBroth
Quantity20 x 5 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
R08130Full specifications
Each20mm x 113mm Tube100 x 10 mLRequest A Quote
Format20mm x 113mm Tube
TypeAgar
DescriptionBHI Broth
Product TypeAgar
Quantity100 x 10 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
R08132Full specifications
Each20mm x 113mm Tube20 x 10 mLRequest A Quote
Format20mm x 113mm Tube
TypeAgar
DescriptionBHI Broth
Product TypeAgar
Quantity20 x 10 mL
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
Unit SizeEach
Showing 10 of 12Show more

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.

Figures