Remel™ BHI Broth (Brain Heart Infusion Broth)
Thermo Scientific™

Remel™ BHI Broth (Brain Heart Infusion Broth)

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.
Have Questions?
Catalog NumberQuantityFormat
R060270100 x 5 mL15mm x 103mm Tube
Catalog number R060270
Price (CLP)
-
Quantity:
100 x 5 mL
Format:
15mm x 103mm Tube

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.
Specifications
Certifications/ComplianceIndustrial Reference: BAM, SMD, SMWW, USDA.
DescriptionBHI Broth
Format15mm x 103mm Tube
Quantity100 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach