Thermo Scientific™

Remel™ Campy Thioglycollate Medium (w/.16% Agar)

Catalog number: R060370
Thermo Scientific™

Remel™ Campy Thioglycollate Medium (w/.16% Agar)

Catalog number: R060370

Thermo Scientific™ Remel Campy Thioglycollate Medium (w/.16% agar, trimethoprim, vancomycin, polymyxin B, cephalothin, amphotericin B) is used as a holding medium for samples prior to inoculation of media for recovery of Campylobacter spp.

 
Catalog Number
R060370
Unit Size
Each
Quantity
100 x 9 mL
Price (USD)
Full specifications
DescriptionCampy Thioglycollate Medium
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Quantity100 x 9 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
20 x 9 mL
Price (USD)
Full specifications
DescriptionCampy Thioglycollate Medium
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Quantity20 x 9 mL
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R060370Full specifications
Each100 x 9 mLRequest A Quote
DescriptionCampy Thioglycollate Medium
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Quantity100 x 9 mL
Unit SizeEach
R060372Full specifications
Each20 x 9 mLRequest A Quote
DescriptionCampy Thioglycollate Medium
Format15mm x 103mm Tube
Product TypeAgar
TypeAgar
Quantity20 x 9 mL
Unit SizeEach
Showing 2 of 2

Thermo Scientific™ Remel Campy Thioglycollate Medium (w/.16% agar, trimethoprim, vancomycin, polymyxin B, cephalothin, amphotericin B) is used as a holding medium for samples prior to inoculation of media for recovery of Campylobacter spp.

Figures