Thermo Scientific™
Remel™ CTA w/1% Arabinose (Cystine Trypticase Agar)
Related applications:
Clinical Microbiology
Thermo Scientific™ Remel CTA with 1% Arabinose (Cystine Trypticase Agar) is used for determination of fermentation reactions by fastidious microorganisms such as Neisseria, pneumococci, streptococci and nonsporeforming anaerobes.
Back to top
Description
Thermo Scientific™ Remel CTA with 1% Arabinose (Cystine Trypticase Agar) is used for determination of fermentation reactions by fastidious microorganisms such as Neisseria, pneumococci, streptococci and nonsporeforming anaerobes.