Thermo Scientific™

D/E (Dey-Engley) Neutralizing Broth

Catalog number: R060770
Thermo Scientific™

D/E (Dey-Engley) Neutralizing Broth

Catalog number: R060770

Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using Thermo Scientific™ Remel™ D/E (Dey-Engley) Neutralizing Broth. This medium was formulated by Engley and Dey in 1970 for neutralizing and testing antiseptics and disinfectants1. It is recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling2.

 
Catalog Number
R060770
Unit Size
Each
dimFormat
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: COMPF.
DescriptionD/E Neutralizing Broth
Format15mm x 103mm Tube
Product TypeMicrobiology Media
Quantity5 mL
Unit SizeEach
Catalog Number
Unit Size
Each
dimFormat
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: COMPF.
DescriptionD/E Neutralizing Broth
Format20mm x 150mm Tube
Product TypeMicrobiology Media
Quantity9 mL
Unit SizeEach
Catalog Number
Unit Size
Each
dimFormat
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: COMPF.
DescriptionD/E Neutralizing Broth
FormatWMB Lug
Product TypeMicrobiology Media
Quantity90 mL
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizedimFormatPrice (USD)
R060770Full specifications
EachRequest A Quote
Certifications/ComplianceIndustrial Reference: COMPF.
DescriptionD/E Neutralizing Broth
Format15mm x 103mm Tube
Product TypeMicrobiology Media
Quantity5 mL
Unit SizeEach
R09334Full specifications
EachRequest A Quote
Certifications/ComplianceIndustrial Reference: COMPF.
DescriptionD/E Neutralizing Broth
Format20mm x 150mm Tube
Product TypeMicrobiology Media
Quantity9 mL
Unit SizeEach
R112160Full specifications
EachRequest A Quote
Certifications/ComplianceIndustrial Reference: COMPF.
DescriptionD/E Neutralizing Broth
FormatWMB Lug
Product TypeMicrobiology Media
Quantity90 mL
Unit SizeEach
Showing 3 of 3

Neutralize chemical disinfectants to distinguish bacteriostatic and bactericidal activity in environmental sampling procedures using D/E (Dey-Engley) Neutralizing Broth. The medium can neutralize a range of chemicals including mercurial, iodine or chlorine preparations, quaternary ammonium compounds, formaldehyde and glutaraldehyde.

  • Ready to Use: Convenience of prepared media.
  • Easy to Identify: As dextrose is utilized by the bacteria the pH indicator changes from a purple to yellow color.
  • Easy to differentiate: Subculture all tubes of D/E Neutralizing Broth to appropriate media such as D/E Agar to observe growth (viable microorganisms).
    • Bacteriostatic substance – Growth from negative D/E Neutralizing Broth tubes.
    • Bactericidal substance – No growth from negative D/E Neutralizing Broth tubes.
  • Recommended in the Compendium of Methods for the Microbiological Examination of Foods by the American Public Health Association (APHA) for environmental sampling.

This medium contains peptone and yeast extract that provide nutrients such as amino acids, peptides, nitrogenous compounds, and B-complex vitamins which are necessary for growth. Dextrose provides energy, lecithin neutralizes quaternary ammonium compounds, polysorbate 80 neutralizes phenolics, thioglycolate neutralizes mercurials, sodium thiosulfate neutralizes iodine and chlorine preparations and sodium bisulfate neutralizes formaldehyde and glutaraldehyde. This medium is suitable for environmental sampling where neutralization of the chemical is important to determine its bactericidal activity.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Not intended for IVD use.
  1. Engley, F.B. and B.P. Dey. 1970. Chemical Specialities Manufacturing Association Proceedings.
  2. Downes, F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th APHA. Washington, D.C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text