Thermo Scientific™

Heart Infusion Broth w/BCP, 1% Mannitol (Brom Cresol Purple), D - 5mL

Catalog number: R061104
Thermo Scientific™

Heart Infusion Broth w/BCP, 1% Mannitol (Brom Cresol Purple), D - 5mL

Catalog number: R061104
Thermo Scientific™ Remel Heart Infusion Broth with BCP, 1% Mannitol (Brom Cresol Purple) for determination of fermentation reactions of microorganisms, especially streptococci and enterococci.
 
Catalog Number
R061104
Unit Size
Each
Price (USD)
Full specifications
Format15mm x 103mm Tube
TypeBroth
DescriptionHeart Infusion Broth w/BCP
Product TypeBroth
Quantity20 x 5 mL
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
R061104Full specifications
EachRequest A Quote
Format15mm x 103mm Tube
TypeBroth
DescriptionHeart Infusion Broth w/BCP
Product TypeBroth
Quantity20 x 5 mL
Unit SizeEach
Showing 1 of 1
Thermo Scientific™ Remel Heart Infusion Broth with BCP, 1% Mannitol (Brom Cresol Purple) for determination of fermentation reactions of microorganisms, especially streptococci and enterococci.

Figures

Frequently asked questions (FAQs)

Citations & References