Thermo Scientific™
Remel™ Letheen Broth
Related applications:
Clinical Microbiology
The Thermo Scientific™ Remel Letheen Broth is for testing quaternary ammonium compounds for antimicrobial activity.
Back to top
Description
The Thermo Scientific™ Remel Letheen Broth is for testing quaternary ammonium compounds for antimicrobial activity.