Search
Search
Selectively and differentially isolate enteric gram-negative bacilli using MacConkey Broth.
| Catalog Number | Format |
|---|---|
| R061336 | D - 5mL |
| R112454 | WMB - 100mL |
| R112456 | PB - 100mL |
Selectively and differentially isolate enteric gram-negative bacilli using Thermo Scientific™ Remel™ MacConkey Broth. MacConkey Broth is a modification of the original bile salt broth first described by MacConkey in 1900, which contained litmus as an indicator and sodium taurocholate to inhibit gram-positive organisms1. In further testing, MacConkey recommended replacing litmus with neutral red2. In 1953, Childs and Allen replaced neutral red with bromcresol purple, a less inhibitory indicator3.
Selectively and differentially isolate enteric gram-negative bacilli using MacConkey Broth. This medium is used for presumptive identification of coliforms. Coliforms are organisms that are present in the environment and in the feces of all warm-blooded animals and humans.
Their presence in drinking water indicates that disease-causing organisms (pathogens) could be in the water system.
This medium contains gelatin peptone that provides amino acids and essential nutrients for bacterial growth. Lactose is a carbon source for energy. Oxgall is a selective agent which inhibits gram-positive organisms. Lactose and bromcresol purple indicator enable the differentiation of lactose fermenting gram-negative bacilli.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.