Search
Search
Cultivate and maintain wide variety of microorganisms with Nutrient Agar.
| Catalog Number | Quantity | Format |
|---|---|---|
| R061572 | 20/Pk. | 15mm x 103mm Tube |
| R061570 | 100/Pk. | 15mm x 103mm Tube |
| R08622 | 20 x 10 mL | 20mm x 113mm Pour Tube |
Cultivate and maintain wide variety of microorganisms with Thermo Scientific™ Remel™ Nutrient Agar. This is a non-selective medium which was published by APHA in 19171, as a standardized medium for the use in the examination of water, wastewater, dairy products and various foods. This medium is also used for the recovery of non-fastidious microorganisms and to cultivate and maintain stock cultures2,3.
Nutrient Agar is a general purpose culture medium used to cultivate and maintain wide variety of microorganisms.
This medium contains beef extract that provides carbohydrates, vitamins, nitrogen compounds, and salts. Gelatin peptone provides nutrients in the form of amino acids and peptides. When used in combination, beef extract and gelatin are found to be as nutritious as meat infusion. Agar is the solidifying agent.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
General References: