OF Medium w/1% Sucrose
Product Image
Thermo Scientific™

OF Medium w/1% Sucrose

Use OF Medium w/1% Sucrose (Hugh-Liefson) to determine oxidative and fermentative metabolism of sucrose by gram-negative bacilli.

Have Questions?
Número de catálogoCantidad
R06199020 x 4 mL
Número de catálogo R061990
Precio (MXN)
-
Cantidad:
20 x 4 mL

Determine oxidative and fermentative metabolism of sucrose by gram-negative bacilli using Thermo Scientific™ Remel™ OF Medium w/1% Sucrose (Hugh-Liefson). Hugh and Leifson first reported that microorganisms which utilize carbohydrates oxidatively produce only a minimal amount of acid1. Oxidative-Fermentative (OF) Medium was developed with a ratio of peptone to carbohydrate that minimized the formation of alkaline by-products and enhanced acid production by oxidative organisms. OF Medium is especially suitable for determining the carbohydrate metabolic pathway of gram-negative bacilli2.

Typical fermentation media contain approximately 2% peptone and 1% carbohydrate. In media containing high concentrations of peptones, oxidative microorganisms produce alkaline by-products in quantities that neutralize the acid produced from carbohydrate metabolism.

OF Medium is a semisolid medium with a low peptone to carbohydrate ratio that contains the pH indicator bromthymol blue. Organisms which metabolize carbohydrate in OF medium produce acid in quantities sufficient to prevent its neutralization by alkaline peptone by-products.

  • Ready to use – Convenience of prepared media
  • Uses 4 mL of 1% sucrose as carbohydrate source
  • Easy to interpret – Color change from green to yellow indicates positive test for acid production.
    When testing for motility, growth diffused away from stab line of inoculation indicates a positive test whereas growth confined to stab line indicates negative test.
    Acid production in the open tube (without mineral oil overlay) indicates oxidative metabolism whereas acid production in both (with and without mineral oil overlay) tubes indicates fermentative metabolism

OF Medium with Sucrose uses 4 mL of 1% sucrose as a fermentable carbohydrate. The medium contains casein peptone as the substrate and bromthymol blue as the pH indicator. Dipotassium phosphate promotes fermentation and also serves as a buffer. Agar allows for detection of motility and aids in even distribution of acid produced at the surface of the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Hugh, R. and E. Liefson. 1953. J. Bacteriol. 66:24.
  2. Gilardi, G.L. 1978. Glucose Non fermenting Gram-Negative Bacteria in Clinical Microbiology. CRC Press Inc., West Palm Beach, FL.
Especificaciones
Certificaciones/conformidadIndustrial Reference: BAM.
DescripciónOF Medium w/1% Sucrose (Hugh-Liefson)
Formato15mm x 103mm Tube
Cantidad20 x 4 mL
Tipo de productoMicrobiology Media
Unit SizeEach