Product Image
Thermo Scientific™
Phenol Red Broth w/1% Fructose
Thermo Scientific™ Remel Phenol Red Broth w/1% Fructose for determination of fermentation reactions by microorganisms.
Have Questions?
Catalog NumberQuantity
R06227220 x 5 mL
Catalog number R062272
Price (MXN)
-
Quantity:
20 x 5 mL
Thermo Scientific™ Remel Phenol Red Broth w/1% Fructose for determination of fermentation reactions by microorganisms.
Specifications
DescriptionPhenol Red Broth w/1% Fructose
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach