Phenol Red Broth w/1% Lactose
Product Image
Thermo Scientific™

Phenol Red Broth w/1% Lactose

Determine fermentation reactions of bacteria using Phenol Red Broth with 1% Lactose.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R06232220 x 5 mL
R06232620 x 7 mL
Catalog number R062322
Price (MXN)
-
Quantity:
20 x 5 mL

Determine fermentation reactions of bacteria with Thermo Scientific™ Remel™ Phenol Red Broth with 1% Lactose.

Vera recommended using casein peptone in fermentation test media because of its high growth-promoting qualities1. It was reported that peptone could be used with phenol red as the pH indicator in fermentation tests.

Phenol Red Broth is a complete basal medium prepared with phenol red as an indicator, which changes the medium to yellow when acid is produced. Carbohydrates may be added to the broth base in a final concentration of 1%. A Phenol Red Broth Base Control tube is used as a negative control for fermentation studies. A Durham tube may be inserted in a tube of Phenol Red Broth with 1% Lactose to allow detection of gas production.

Can be used in fermentation studies of pure culture of many species of bacteria due to high growth promoting qualities of casein peptone.

  • Recommended by Association of Official Analytical Collaboration (AOAC) International and the American Public Health Association (APHA)2-5.
  • Easy to interpret because of fermentation reaction and gas production. Yellow color is observed when there is acid production and red-orange color is observed when there is no fermentation. Bubbles are produced in the Durham tube when gas is produced and no bubbles is produced in the Durham tube when there is no gas production.

Phenol Red Broth with 1% Lactose contains casein peptone that has high growth promoting qualities. Phenol red is the indicator which helps to determine the reaction by color change.

This medium also contains Sodium Chloride, Phenol Red, Demineralized Water and Lactose. Sodium Chloride supplies essential electrolytes and maintains osmotic equilibrium of the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Vera, H.D. 1950. Am. J. Public Health. 40:1267-1272.
  2. Food and Drug Administration. 2001. Bacteriological Analytical Manual Online. Chapter 14. Authors: E.J. Rhodehamel and S.M. Harmon. AOAC International, Gaithersburg, MD.
  3. Eaton, A.D., L.S. Clesceri, E.W. Rice, and A.E. Greenberg. 2005. Standard Methods for the Examination of Water and Wastewater. 21st APHA, Washington, D.C.
  4. Wehr, H.M. and J.F. Frank. 2004. Standard Methods for the Examination of Dairy Products. 17th APHA, Washington, D.C.
  5. Downes, F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th APHA, Washington, D.C.
Specifications
DescriptionPhenol Red Broth w/1% Lactose
Format15mm x 103mm Tube
Quantity20 x 5 mL
Product TypeMicrobiology Media
Unit SizeEach