Product Image
Thermo Scientific™

Phenol Red Broth w/1% Salicin

Determine fermentation reactions of bacteria using Phenol Red Broth w/1% Salicin.

Have Questions?
Número de catálogoDescripción
R062402Caldo rojo fenol con 1 % de salicina
Número de catálogo R062402
Precio (MXN)
-
Descripción:
Caldo rojo fenol con 1 % de salicina

Determine fermentation reactions of bacteria with Thermo Scientific™ Remel™ Phenol Red Broth w/1% Salicin.

Vera recommended using casein peptone in fermentation test media because of its high growth-promoting qualities1. It was reported that peptone could be used with phenol red as the pH indicator in fermentation tests.

Phenol Red Broth is a complete basal medium prepared with phenol red as an indicator, which changes the medium to yellow when acid is produced. Carbohydrates may be added to the broth base in a final concentration of 1%. A Phenol Red Broth Base Control tube is used as a negative control for fermentation studies. A Durham tube may be inserted in a tube of Phenol Red Broth w/1% Salicin to allow detection of gas production.

  • Can be used in fermentation studies of pure culture of many species of bacteria due to high growth promoting qualities of casein peptone.
  • Recommended by Association of Official Analytical Collaboration AOAC International and the American Public Health Association (APHA)2-5.
  • Easy to interpret because of fermentation reaction and gas production. Yellow color is observed when there is acid production and red-orange color is observed when there is no fermentation. Bubbles are produced in the Durham tube when gas is produced and no bubbles is produced in the Durham tube when there is no gas production.

Phenol Red Broth w/1% Salicin contains casein peptone that has high growth promoting qualities. Phenol red is the indicator which helps to determine the reaction by color change.

This medium also contains Sodium Chloride, Phenol Red, Demineralized Water and Salicin. Sodium Chloride supplies essential electrolytes and maintains osmotic equilibrium of the medium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

  1. Vera, H.D. 1950. Am. J. Public Health. 40:1267-1272.
  2. Food and Drug Administration. 2001. Bacteriological Analytical Manual Online. Chapter 14. Authors: E.J. Rhodehamel and S.M. Harmon. AOAC International, Gaithersburg, MD.
  3. Eaton, A.D., L.S. Clesceri, E.W. Rice, and A.E. Greenberg. 2005. Standard Methods for the Examination of Water and Wastewater. 21st APHA, Washington, D.C.
  4. Wehr, H.M. and J.F. Frank. 2004. Standard Methods for the Examination of Dairy Products. 17th APHA, Washington, D.C.
  5. Downes, F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th APHA, Washington, D.C.
Especificaciones
DescripciónCaldo rojo fenol con 1 % de salicina
Formato15mm x 103mm Tube
Cantidad20 x 5 mL
Tipo de productoMicrobiology Media
Unit SizeEach